Skip to Content
Merck
β-Endorphin rat ≥97% (HPLC)

β-Endorphin rat

β-Endorphin rat ≥97% (HPLC)
Synonym(s):
β-Lipotropin 61-91
Empirical Formula (Hill Notation):
C157H254N42O44S
CAS Number:
Molecular Weight:
3466.02

Products

Product Category
Brand
Formula Weight
Gene Alias
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing