Skip to Content
Merck
vibrant-m

Azathioprine impurity G

vibrant-m
Synonym(s):
6-[(1-Methyl-4-nitro-1H-imidazol-5-yl)sulfanyl]-7H-purin-2-amine
Empirical Formula (Hill Notation):
C9H8N8O2S
CAS Number:
Molecular Weight:
292.28

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing