Aripiprazole Related Compound F
Synonym(s):
7-[4-[4-(2,3-Dichlorophenyl)-1-oxidopiperazin-1-yl]butoxy]-3,4-dihydroquinolein-2(1H)-one, 4-(2,3-Dichlorophenyl)-1-[4-(2-oxo-1,2,3,4-tetrahydroquinolin-7-yloxy)butyl]piperazin 1-oxide
Empirical Formula (Hill Notation):
C23H27Cl2N3O3
CAS Number:
Molecular Weight:
464.38
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2