Skip to Content
Merck
Arg-Gly-Asp-Ser-Pro-Ala-Ser-Ser-Lys-Pro ≥97% (HPLC)

Arg-Gly-Asp-Ser-Pro-Ala-Ser-Ser-Lys-Pro

Arg-Gly-Asp-Ser-Pro-Ala-Ser-Ser-Lys-Pro ≥97% (HPLC)
Empirical Formula (Hill Notation):
C40H68N14O16
CAS Number:
Molecular Weight:
1001.05

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing