Anti-Retinoblastoma, C-Terminal antibody produced in rabbit
Synonym(s):
P105-RB, PP105, PP110, RB-1, RB1
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing