Anti-Pyk2 antibody produced in rabbit
Synonym(s):
Anti-CAK-β, Anti-Proline rich Kinase 2, CADTK, CAK β, Calcium-dependent tyrosine kinase, Cell adhesion kinase β, FADK 2
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2