Skip to Content
Merck
Anti-MAP2 (2a+2b) antibody, Mouse monoclonal ~2 mg/mL, clone AP-20, purified from hybridoma cell culture

Anti-MAP2 (2a+2b) antibody, Mouse monoclonal

Anti-MAP2 (2a+2b) antibody, Mouse monoclonal ~2 mg/mL, clone AP-20, purified from hybridoma cell culture
Synonym(s):
Anti-Microtubule Associated Protein 2

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique