
Anti-KHSRP antibody produced in rabbit

Synonym(s):
Anti-FBP2, Anti-FUBP2, Anti-KH-type splicing regulatory protein, Anti-KSRP, Anti-MGC99676
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing