Anti-Human IgG3−FITC antibody, Mouse monoclonal
Synonym(s):
Monoclonal Anti-Human IgG3
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing