Skip to Content
Merck
Anti-HOXA3 antibody produced in rabbit ~1.0 mL/mL, affinity isolated antibody

Anti-HOXA3 antibody produced in rabbit

Anti-HOXA3 antibody produced in rabbit ~1.0 mL/mL, affinity isolated antibody
Synonym(s):
Anti-HOX1, Anti-HOX1E, Anti-Homeobox A3, Anti-MGC10155

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing