
Anti-GPKOW antibody produced in rabbit

Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing