
Anti-COX2 antibody produced in rabbit

Synonym(s):
COX2, CYCLOOXYGENASE 2, GLUCOCORTICOID-REGULATED INFLAMMATORY PROSTAGLANDIN G/H SYNTHASE, GRIPGHS, PGHS2
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3