Anti-c-Myc Tag Antibody
Synonym(s):
Anti-c-Myc, Anti-Cellular myelocytomatosis oncogene, Anti-Myc proto-oncogene protein, Myc proto-oncogene protein, c-myc, Anti- MRTL, Anti- bHLHe39, Anti-Myc protein, Anti-cMyc, Anti-v-Myc myelocytomatosis viral oncogene homolog (avian), Anti-bHLHe39 antibody produced in rabbit, Anti-c-Myc antibody produced in rabbit, Anti-v-myc myelocytomatosis viral oncogene homolog (avian) antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-20 of 21
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2