Alfadex
Synonym(s):
α-Cyclodextrin, α-Schardinger dextrin, Cyclohexaamylose, Cyclomaltohexaose
Empirical Formula (Hill Notation):
C36H60O30
CAS Number:
Molecular Weight:
972.84
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing