
Acetamiprid-N-desmethyl

Synonym(s):
N-(6-Chloro-3-pyridylmethyl)-N′-cyano-acetamidine
Empirical Formula (Hill Notation):
C9H9ClN4
CAS Number:
Molecular Weight:
208.65
Products
Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing