
Acamprosate calcium

Synonym(s):
3-(Acetylamino)-1-propanesulfonic acid calcium salt (2:1), Acamprosate calcium
Empirical Formula (Hill Notation):
C5H10Ca0.5NO4S
CAS Number:
Molecular Weight:
200.24
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-3 of 3