Skip to Content
Merck
6-Chloro-2,4-diamino-1,3,5-triazine-13C3 VETRANAL®, analytical standard

6-Chloro-2,4-diamino-1,3,5-triazine-13C3

6-Chloro-2,4-diamino-1,3,5-triazine-13C3 VETRANAL®, analytical standard
Empirical Formula (Hill Notation):
13C3H4ClN5
CAS Number:
Molecular Weight:
148.53

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Isotope
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique