5-Bromo-4-chloro-3-indolyl α-D-galactopyranoside
Synonym(s):
X-α-Gal
Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing