Skip to Content
Merck
5,5′,6,6′-Tetramethyl-3,3′-di-tert-butyl-1,1′-biphenyl-2,2′-diol 97%

5,5′,6,6′-Tetramethyl-3,3′-di-tert-butyl-1,1′-biphenyl-2,2′-diol

5,5′,6,6′-Tetramethyl-3,3′-di-tert-butyl-1,1′-biphenyl-2,2′-diol 97%
Empirical Formula (Hill Notation):
C24H34O2
CAS Number:
Molecular Weight:
354.53

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing