Skip to Content
Merck
4-Methylumbelliferyl N-acetyl-β-D-galactosaminide ≥98% (HPLC)

4-Methylumbelliferyl N-acetyl-β-D-galactosaminide

4-Methylumbelliferyl N-acetyl-β-D-galactosaminide ≥98% (HPLC)
Synonym(s):
4-Methylumbelliferyl 2-acetamido-2-deoxy-β-D-galactopyranoside
Empirical Formula (Hill Notation):
C18H21NO8
CAS Number:
Molecular Weight:
379.36
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing