Skip to Content
Merck
4-Hydroxybenzhydrazide ≥97%

4-Hydroxybenzhydrazide

4-Hydroxybenzhydrazide ≥97%
Synonym(s):
4-Hydroxybenzoic hydrazide, 4-Hydroxybenzoylhydrazine, p-Hydroxybenzoic acid hydrazide, 4-Hydroxybenzhydrazide
Linear Formula:
HOC6H4CONHNH2
CAS Number:
Molecular Weight:
152.15

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing