![4-amino-2-methyl-pyrimidine-5-carbonitrile AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/378/692/20c14c6c-bad8-4971-8860-f4911c875769/640/20c14c6c-bad8-4971-8860-f4911c875769.png)
4-amino-2-methyl-pyrimidine-5-carbonitrile
![4-amino-2-methyl-pyrimidine-5-carbonitrile AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/378/692/20c14c6c-bad8-4971-8860-f4911c875769/640/20c14c6c-bad8-4971-8860-f4911c875769.png)
Empirical Formula (Hill Notation):
C6H6N4
CAS Number:
Molecular Weight:
134.14
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing