Skip to Content
Merck
2-Thiopheneacetonitrile 97%

2-Thiopheneacetonitrile

2-Thiopheneacetonitrile 97%
Synonym(s):
2-Thienylacetonitrile
Empirical Formula (Hill Notation):
C6H5NS
CAS Number:
Molecular Weight:
123.18
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing