Skip to Content
Merck
2-Phenylindole technical grade, 95%

2-Phenylindole

2-Phenylindole technical grade, 95%
Synonym(s):
NSC 15776, Stabilizer I
Empirical Formula (Hill Notation):
C14H11N
CAS Number:
Molecular Weight:
193.24
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing