Skip to Content
Merck
2′-Deoxyadenosine 5′-triphosphate disodium salt ≥97%

2′-Deoxyadenosine 5′-triphosphate disodium salt

2′-Deoxyadenosine 5′-triphosphate disodium salt ≥97%
Synonym(s):
dATP-Na2
Empirical Formula (Hill Notation):
C10H14N5Na2O12P3
CAS Number:
Molecular Weight:
535.15
EC Number:

Products

Brand
Biological Source
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing