2-Chlorothioxanthone
Synonym(s):
2-Chlorothioxanthen-9-one
Empirical Formula (Hill Notation):
C13H7ClOS
CAS Number:
Molecular Weight:
246.71
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing