
2-Chloro-6-methylpyrazine

Empirical Formula (Hill Notation):
C5H5ClN2
CAS Number:
Molecular Weight:
128.56
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing