![2-Butyne-1,4-diol 99%](/deepweb/assets/sigmaaldrich/product/structures/260/147/d54b4951-c827-4423-9d4c-471af5d003d3/640/d54b4951-c827-4423-9d4c-471af5d003d3.png)
2-Butyne-1,4-diol
![2-Butyne-1,4-diol 99%](/deepweb/assets/sigmaaldrich/product/structures/260/147/d54b4951-c827-4423-9d4c-471af5d003d3/640/d54b4951-c827-4423-9d4c-471af5d003d3.png)
Synonym(s):
2-Butyne-1,4-diol
Linear Formula:
HOCH2C≡CCH2OH
CAS Number:
Molecular Weight:
86.09
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing