Skip to Content
Merck
2-Amino-4-chloropyrimidine 97%

2-Amino-4-chloropyrimidine

2-Amino-4-chloropyrimidine 97%
Empirical Formula (Hill Notation):
C4H4ClN3
CAS Number:
Molecular Weight:
129.55

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing