![2,2-Difluoro-1-methylcyclopropylamine hydrochloride AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/152/864/3b7356e8-0ede-4c3a-93b9-b9693d878bad/640/3b7356e8-0ede-4c3a-93b9-b9693d878bad.png)
2,2-Difluoro-1-methylcyclopropylamine hydrochloride
![2,2-Difluoro-1-methylcyclopropylamine hydrochloride AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/152/864/3b7356e8-0ede-4c3a-93b9-b9693d878bad/640/3b7356e8-0ede-4c3a-93b9-b9693d878bad.png)
Synonym(s):
1-Amino-2,2-difluoro-1-methylcyclopropane hydrochloride, 2,2-Difluoro-1-methylcyclopropanamine hydrochloride
Empirical Formula (Hill Notation):
C4H7F2N·HCl
CAS Number:
Molecular Weight:
143.56
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing