Skip to Content
ID
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1
H
,1
H
,2
H
,2
H
-Perfluorohexan-1-ol
Synonym(s):
3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol
Linear Formula:
CF
3
(CF
2
)
3
(CH
2
)
2
OH
CAS Number:
2043-47-2
Molecular Weight:
264.09
EC Number:
218-050-9
Products
Product Category
building blocks (1)
Boiling Point (°C)
Formula Weight
Purity
Application
Markush Class
Markush Group
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
532770
1
H
,1
H
,2
H
,2
H
-Perfluorohexan-1-ol,
97%
Expand
View Pricing