Skip to Content
Merck
1-Deoxy-D-xylulose-5-phosphate sodium salt ≥99.0% (TLC)

1-Deoxy-D-xylulose-5-phosphate sodium salt

1-Deoxy-D-xylulose-5-phosphate sodium salt ≥99.0% (TLC)
Synonym(s):
DXP sodium salt
Empirical Formula (Hill Notation):
C5H11O7P · xNa+
CAS Number:
Molecular Weight:
214.11 (free acid basis)

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing