
1-(α,α,α-Trifluoro-m-tolyl)piperazine hydrochloride

Synonym(s):
N-[3-(Trifluoromethyl)phenyl]piperazine hydrochloride, TFMPP hydrochloride
Empirical Formula (Hill Notation):
C11H13F3N2 · HCl
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Research Area
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing