
(−)-1-(9-Fluorenyl)ethyl chloroformate solution

Synonym(s):
(−)-FLEC
Empirical Formula (Hill Notation):
C16H13ClO2
CAS Number:
Molecular Weight:
272.73
Products
Product Category
Brand
Formula Weight
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing