1,6-Hexanediol
Synonym(s):
Hexamethylene glycol, 1,6-Hexanediol
Linear Formula:
HO(CH2)6OH
CAS Number:
Molecular Weight:
118.17
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-3 of 3
Compare
Product Number
Description
Pricing