![1,4-Dichlorobutane 99%](/deepweb/assets/sigmaaldrich/product/structures/113/442/8616c6dd-0dfb-418b-a640-899d76511da6/640/8616c6dd-0dfb-418b-a640-899d76511da6.png)
1,4-Dichlorobutane
![1,4-Dichlorobutane 99%](/deepweb/assets/sigmaaldrich/product/structures/113/442/8616c6dd-0dfb-418b-a640-899d76511da6/640/8616c6dd-0dfb-418b-a640-899d76511da6.png)
Synonym(s):
Tetramethylene dichloride, 1,4-Dichlorobutane
Linear Formula:
Cl(CH2)4Cl
CAS Number:
Molecular Weight:
127.01
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing