Skip to Content
Merck
1,4,5,8-Naphthalenetetracarboxylic dianhydride

1,4,5,8-Naphthalenetetracarboxylic dianhydride

1,4,5,8-Naphthalenetetracarboxylic dianhydride
Synonym(s):
NTCDA
Empirical Formula (Hill Notation):
C14H4O6
CAS Number:
Molecular Weight:
268.18
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing