Skip to Content
Merck
1,3,5-Phenyltriboronic acid, tris(pinacol) ester ≥97%

1,3,5-Phenyltriboronic acid, tris(pinacol) ester

1,3,5-Phenyltriboronic acid, tris(pinacol) ester ≥97%
Empirical Formula (Hill Notation):
C24H39B3O6
CAS Number:
Molecular Weight:
456.00

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing