1-[3-(4-tert-Butylbenzoyl)propyl]-4-hydroxypiperidine
Synonym(s):
1-[4-(1,1-Dimethylethyl)phenyl]-4-(4-hydroxypiperidin-1-yl)butan-1-one
Empirical Formula (Hill Notation):
C19H29NO2
CAS Number:
Molecular Weight:
303.44
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing