![1,1,2-Trimethyl-3-(4-sulfobutyl)benz[e]indolium, inner salt Dye content 90 %](/deepweb/assets/sigmaaldrich/product/structures/878/047/5e72fb07-3f29-4adf-877d-b176efdb3f87/640/5e72fb07-3f29-4adf-877d-b176efdb3f87.png)
1,1,2-Trimethyl-3-(4-sulfobutyl)benz[e]indolium, inner salt
![1,1,2-Trimethyl-3-(4-sulfobutyl)benz[e]indolium, inner salt Dye content 90 %](/deepweb/assets/sigmaaldrich/product/structures/878/047/5e72fb07-3f29-4adf-877d-b176efdb3f87/640/5e72fb07-3f29-4adf-877d-b176efdb3f87.png)
Synonym(s):
1,1,2-Trimethyl-3-(4-sulfobutyl)-1H-benz[e]indolium hydroxide, inner salt, 1-(4-Sulfobutyl)-2,3,3-trimethyl-4,5-benzindolium, inner salt
Empirical Formula (Hill Notation):
C19H23NO3S
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing