Product Name
6-Fluoro-2-naphthoic acid 97%
Product Number
638862
Product Brand
aldrich
CAS Number
5043-01-6
Molecular Weight
190.17
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Light Yellow and Faint Beige to Light Beige and Faint Brown to Light Brown |
APPEARANCE (FORM) | Powder or Crystals |
TITRATION (T) NAOH 0.1M | 96.5 - 103.5 % |
PURITY (GC AREA %) | ≥ 96.5 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
RECOMMENDED RETEST PERIOD | 72 MONTHS |