Skip to Content
Merck
All Photos(1)

Key Documents

Z637262

Edisonite Super universal detergent

weight 5 kg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800

description

neutral detergent

manufacturer/tradename

BRAND 44365

weight

5 kg

Looking for similar products? Visit Product Comparison Guide

General description

Neutral detergent. Powder compound for immersion and ultrasonic bath. Ideal for removal of natural fats and oils, protein and blood. Edisonite SUPER is recommended for optical glass, for plastics sensitive to alkalis and rubbers.

Other Notes

BR44365 (India)

Legal Information

Edisonite is a trademark of Merz GmbH & Co. KG

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service