Skip to Content
Merck
All Photos(1)

Key Documents

C9051

Sigma-Aldrich

Cobalt naphthenate

solution

Synonym(s):

Co-Naphthenate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002

form

solution

storage temp.

2-8°C

InChI

1S/2C10H18O2.Co/c2*1-2-8-3-4-9(7-8)5-6-10(11)12;/h2*8-9H,2-7H2,1H3,(H,11,12);/q;;+2/p-2

InChI key

XSWKLHINRKWMTD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Accelerator for polyester embedding media.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service