Skip to Content
Merck
All Photos(1)

Documents

59761

Supelco

Bisabolol oxide A

analytical standard

Synonym(s):

(3S,6S)-Tetrahydro-2,2,6-trimethyl-6-[(1S)-4-methyl-3-cyclohexen-1-yl]-2H-pyran-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O2
CAS Number:
Molecular Weight:
238.37
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

[H][C@@]1(CCC(C)=CC1)[C@]2(C)CC[C@H](O)C(C)(C)O2

InChI

1S/C15H26O2/c1-11-5-7-12(8-6-11)15(4)10-9-13(16)14(2,3)17-15/h5,12-13,16H,6-10H2,1-4H3/t12-,13+,15+/m1/s1

InChI key

WJHRAVIQWFQMKF-IPYPFGDCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ikuko Ogata et al.
Archives of toxicology, 84(1), 45-52 (2009-10-17)
German chamomile (Matricaria recutita L.), one of the popular ingredients in herbal teas, has been traditionally used for medicinal purposes. Bisabololoxide A (BSBO) is one of the main constituents in this herb. BSBO is supposed to be principle in some
Ikuko Ogata-Ikeda et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 18(5), 362-365 (2010-09-25)
German chamomile (Matricaria recutita L.) is a popular ingredient in herbal teas. In previous study, micromolar bisabololoxide A, one of main constituents in German chamomile, exerted cytotoxic action on rat thymocyte, a normal non-proliferative cell. This result prompted us to
Cristina Avonto et al.
Journal of natural products, 76(10), 1848-1853 (2013-09-28)
German chamomile (Matricaria chamomilla) is one of the most popular medicinal plants used in Western herbal medicine. Among the various phytochemicals present in the essential oil of the flowers of German chamomile, bisabolol and its oxidized metabolites are considered as
Eri Fukunaga et al.
Phytotherapy research : PTR, 28(5), 685-691 (2013-07-23)
Bisabololoxide A (BSBO), main constituents in German chamomile extract, is responsible for antipruritic effect. In previous study, the incubation with 30-100 μM BSBO for 24 h exerted cytotoxic and proapoptotic effects on rat thymocytes. To further characterize BSBO cytotoxicity, the effect on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service