Skip to Content
Merck

1.07813

SAFC

Tin(II) chloride dihydrate

EMPROVE® ESSENTIAL, Ph. Eur., BP, JPE

Pharma Manufacturing

Synonym(s):

Tin(II) chloride dihydrate, Tin dichloride, dihydrate, Stannous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Sn · 2H2O
CAS Number:
Molecular Weight:
225.65
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-868-0

Agency

BP
JPE
Ph. Eur.

Quality Level

product line

EMPROVE® ESSENTIAL

form

solid

pH

1-2 (20 °C, 100 g/L in H2O)

bp

623 °C/1013 hPa (anhydrous substance)

mp

38 °C (anhydrous substance)

density

2.71 g/cm3 at 20 °C

application(s)

pharmaceutical

storage temp.

15-25°C

InChI

1S/2ClH.2H2O.Sn/h2*1H;2*1H2;/q;;;;+2/p-2

InChI key

FWPIDFUJEMBDLS-UHFFFAOYSA-L

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service