Skip to Content
Merck
All Photos(1)

Key Documents

KEY366276598

Key Organics / BIONET

1-benzyl-5,5-dimethylimidazolidin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
204.27
MDL number:

mp

142 - 146

Iupac Name

1-benzyl-5,5-dimethyl-2-imidazolidinone

Inchi Code

1S/C12H16N2O/c1-12(2)9-13-11(15)14(12)8-10-6-4-3-5-7-10/h3-7H,8-9H2,1-2H3,(H,13,15)

InChI key

XSKHLLQHJQAERZ-UHFFFAOYSA-N

Purity

97%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/DG-0042

Melting Point

142 - 146

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/DG-0042


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service