CH4312834638
ChemBridge Corporation
tert-butyl 6-(aminomethyl)-1,4-oxazepane-4-carboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
tert-butyl 6-(aminomethyl)-1,4-oxazepane-4-carboxylate
Inchi Code
1S/C11H22N2O3/c1-11(2,3)16-10(14)13-4-5-15-8-9(6-12)7-13/h9H,4-8,12H2,1-3H3
InChI key
CRSZXGHDSAVXGB-UHFFFAOYSA-N
storage temp.
-20C
Purity
90%
Country of Origin
KZ
Shipping Temperature
RT
Physical Form
liquid
Product Link
https://www.hit2lead.com/building-blocks/4038338
MSDS
https://www.hit2lead.com/sds/4038338_sds.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service