Skip to Content
Merck
All Photos(1)

Key Documents

ATEH97A4DC07

AstaTech, Inc.

4-FLUORO-1-METHYLPIPERIDINE-4-CARBOXYLIC ACID

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
161.18
MDL number:

Iupac Name

4-fluoro-1-methyl-4-piperidinecarboxylic acid

Inchi Code

1S/C7H12FNO2/c1-9-4-2-7(8,3-5-9)6(10)11/h2-5H2,1H3,(H,10,11)

InChI key

FQCVRGQFNJITLF-UHFFFAOYSA-N

Purity

93%

Country of Origin

CN

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=178188

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service