Skip to Content
Merck
All Photos(1)

Key Documents

ATE514316297

AstaTech, Inc.

1-(2,6-BIS(BENZYLOXY)PHENYL)ETHANONE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
332.4
MDL number:

Iupac Name

1-[2,6-bis(benzyloxy)phenyl]ethanone

Inchi Code

1S/C22H20O3/c1-17(23)22-20(24-15-18-9-4-2-5-10-18)13-8-14-21(22)25-16-19-11-6-3-7-12-19/h2-14H,15-16H2,1H3

InChI key

ZQJARFAIOBJZSO-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=80856

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=W11652


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service