Skip to Content
Merck
All Photos(1)

Key Documents

ATE341033218

AstaTech, Inc.

2-(3,5-DICHLOROPHENYL) MORPHOLINE OXALATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
322.14
MDL number:

Iupac Name

2-(3,5-dichlorophenyl)morpholine oxalate

Inchi Code

1S/C10H11Cl2NO.C2H2O4/c11-8-3-7(4-9(12)5-8)10-6-13-1-2-14-10;3-1(4)2(5)6/h3-5,10,13H,1-2,6H2;(H,3,4)(H,5,6)

InChI key

SCMJVEVHDSRJCV-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=9869

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=AR1109


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service